Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014648.3 | NP_055463.1 | 9 | 1174 | ||
XM_005247914.3 | XP_005247971.1 | 9 | 973 | |||
XM_005247915.3 | XP_005247972.1 | 9 | 943 | |||
XM_005247916.3 | XP_005247973.1 | 10 | 1115 | |||
XM_005247917.3 | XP_005247974.1 | 9 | 904 | |||
XM_005247918.3 | XP_005247975.1 | 9 | 1391 | |||
XM_005247919.3 | XP_005247976.1 | 9 | 1143 | |||
XM_017007540.1 | XP_016863029.1 | 9 | 1020 | |||
GenBank mRNA | AB014575.1 | - | 9 | 881 | ||
AK023138.1 | - | 9 | 942 | |||
AK023389.1 | - | 9 | 983 | |||
AY227651.1 | - | 9 | 920 | |||
AY227654.1 | - | 9 | 959 | |||
BC039018.1 | - | 9 | 935 | |||
BC056674.1 | - | 9 | 946 | |||
BC063882.1 | - | 9 | 927 | 30387514 |