Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JOSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014876.5 | NP_055691.1 | 4 | 1071 | ||
XM_005261876.2 | XP_005261933.1 | 5 | 1378 | |||
XM_005261877.2 | XP_005261934.1 | 5 | 1274 | |||
XM_005261878.2 | XP_005261935.1 | 5 | 1302 | |||
XM_005261879.2 | XP_005261936.1 | 5 | 1397 | |||
GenBank mRNA | AK291827.1 | - | 4 | 864 | ||
BC015026.2 | - | 4 | 905 | 3921785 | ||
CR456473.1 | - | 4 | 750 | |||
D31884.1 | - | 4 | 883 |