Search Thermo Fisher Scientific
Gene Symbol: CLIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199570.1 | NP_001186499.1 | 6 | 1144 | ||
NM_015526.2 | NP_056341.1 | 7 | 1060 | |||
GenBank mRNA | AJ427922.1 | - | 7 | 1038 | ||
AK056445.1 | - | 5 | 544 | |||
AK090732.1 | - | 7 | 1063 | |||
AK092354.1 | - | 3 | 307 | |||
AK094738.1 | - | 7 | 1058 | |||
AK289509.1 | - | 6 | 1112 | |||
AK294748.1 | - | 7 | 931 | |||
BC013116.1 | - | 7 | 972 | 4214441 | ||
BC014486.1 | - | 4 | 503 | |||
BC128090.1 | - | 3 | 312 |