Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIBC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015653.4 | NP_056468.3 | 3 | 528 | ||
XM_005261524.4 | XP_005261581.1 | 3 | 353 | |||
XM_017028766.1 | XP_016884255.1 | 3 | 450 | |||
GenBank mRNA | AK098586.1 | - | 3 | 528 | ||
AK225722.1 | - | 3 | 509 | |||
AL442116.1 | - | 3 | 479 | |||
BC003024.1 | - | 3 | 490 | 2822570 | ||
BC009904.2 | - | 3 | 490 | 2822570 | ||
CR456438.1 | - | 1 | 122 | |||
CR457398.1 | - | 1 | 113 |