Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129884.2 | NP_001123356.1 | 5, 6 | 860 | ||
NM_001243725.1 | NP_001230654.1 | 4, 5 | 968 | |||
NM_001243726.1 | NP_001230655.1 | 4, 5 | 752 | |||
NM_001243727.2 | NP_001230656.1 | 3, 4 | 586 | |||
NM_001278195.1 | NP_001265124.1 | 5, 6 | 900 | |||
NM_015926.5 | NP_057010.1 | 4, 5 | 808 | |||
NR_024012.2 | 3, 4 | 516 | ||||
NR_103462.1 | 3, 4 | 479 | ||||
NR_103463.1 | 3, 4 | 448 | ||||
XM_006713195.3 | XP_006713258.1 | 4, 5 | 748 | |||
XM_006713197.3 | XP_006713260.1 | 4, 5 | 715 | |||
XM_006713198.2 | XP_006713261.1 | 4, 5 | 783 | |||
XM_011533805.2 | XP_011532107.1 | 4, 5 | 1463 | |||
XM_017006573.1 | XP_016862062.1 | 4, 5 | 968 | |||
GenBank mRNA | AF072733.1 | - | 3, 4 | 706 | ||
AK023786.1 | - | 1 | 2317 | |||
AK023997.1 | - | 4, 5 | 751 | |||
AK222938.1 | - | 4, 5 | 720 | |||
AK222949.1 | - | 4, 5 | 720 | |||
AL832608.1 | - | 2, 3 | 3357 | |||
AY358972.1 | - | 3, 4 | 707 | |||
BC001592.1 | - | 4, 5 | 730 | |||
BC008742.2 | - | 4, 5 | 968 | 3346733 | ||
BG326586.1 | - | 3, 4 | 503 | |||
BG772477.1 | - | 3, 4 | 521 | |||
CB142778.1 | - | 3, 4 | 480 |