Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PEX5L SIRNA Assays ›
Gene Symbol:
PEX5L
Gene Name:
peroxisomal biogenesis factor 5 like
Gene Aliases:
PEX5R, PEX5RP, PXR2, PXR2B, TRIP8b
Chromosome Location:
Chr.3: 179794958 - 180037053 on Build GRCh38
UniGene ID:
Hs.235331
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124976
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PEX5L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256750.1
NP_001243679.1
8
1214
NM_001256751.1
NP_001243680.1
7
1148
NM_001256752.1
NP_001243681.1
7
1115
NM_001256753.1
NP_001243682.1
6
1043
NM_001256754.1
NP_001243683.1
8
950
NM_001256755.1
NP_001243684.1
7
755
NM_001256756.1
NP_001243685.1
7
845
NM_016559.2
NP_057643.1
8
1220
XM_011512882.2
XP_011511184.1
9
1832
XM_011512884.2
XP_011511186.1
9
1828
XM_011512885.2
XP_011511187.1
9
1243
XM_011512886.2
XP_011511188.1
8
1049
XM_011512887.2
XP_011511189.1
7
611
XM_011512888.2
XP_011511190.1
7
911
XM_011512891.2
XP_011511193.1
6
1408
XM_011512892.2
XP_011511194.1
5
1214
XM_017006601.1
XP_016862090.1
8
1640
XM_017006602.1
XP_016862091.1
8
1045
XM_017006603.1
XP_016862092.1
8
1636
XM_017006604.1
XP_016862093.1
6
2226
XM_017006605.1
XP_016862094.1
6
911
XM_017006606.1
XP_016862095.1
8
1717
XM_017006607.1
XP_016862096.1
7
2414
XM_017006608.1
XP_016862097.1
7
1064
XM_017006609.1
XP_016862098.1
7
1557
XM_017006610.1
XP_016862099.1
6
992
XM_017006611.1
XP_016862100.1
6
1369
GenBank mRNA
AB032592.1
-
8
931
AB032593.1
-
8
896
AK294227.1
-
5
445
AK294501.1
-
6
918
AK295349.1
-
7
613
AK295386.1
-
7
845
AK299633.1
-
7
1021
AK303716.1
-
8
941
AK314283.1
-
8
1098
BC036183.1
-
7
999
4793921
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into peroxisome matrix, docking
regulation of cAMP-mediated signaling
peroxisome targeting sequence binding
small GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0