Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIDT1 SIRNA Assays ›
Gene Symbol:
SIDT1
Gene Name:
SID1 transmembrane family member 1
Gene Aliases:
SID-1, SID1
Chromosome Location:
Chr.3: 113531774 - 113629579 on Build GRCh38
UniGene ID:
Hs.591291
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125012
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIDT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308350.1
NP_001295279.1
21, 22
2706
NM_001322294.1
NP_001309223.1
21, 22
2709
NM_001322295.1
NP_001309224.1
20, 21
2622
NM_001322296.1
NP_001309225.1
19, 20
2520
NM_001322297.1
NP_001309226.1
20, 21
2584
NM_001322298.1
NP_001309227.1
20, 21
2683
NM_001322299.1
NP_001309228.1
20, 21
2710
NM_001322300.1
NP_001309229.1
20, 21
2603
NM_017699.2
NP_060169.2
20, 21
2691
NR_136280.1
21, 22
2712
NR_136281.1
20, 21
2651
NR_136282.1
20, 21
2632
NR_136283.1
19, 20
2495
NR_136284.1
17, 18
2402
NR_136285.1
16, 17
2265
XM_011512939.2
XP_011511241.1
20, 21
2654
XM_011512941.2
XP_011511243.1
21, 22
2666
XM_017006664.1
XP_016862153.1
18, 19
2582
XM_017006665.1
XP_016862154.1
19, 20
3097
XM_017006666.1
XP_016862155.1
18, 19
1574
XM_017006667.1
XP_016862156.1
16, 17
1441
XM_017006668.1
XP_016862157.1
9, 10
898
GenBank mRNA
AK295938.1
-
10, 11
894
AK303385.1
-
6, 7
625
BC117222.1
-
21, 22
2094
40125773
BC143776.1
-
19, 20
2020
BC143778.1
-
20, 21
2098
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
dsRNA transport
RNA transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0