Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANSL3 SIRNA Assays ›
Gene Symbol:
KANSL3
Gene Name:
KAT8 regulatory NSL complex subunit 3
Gene Aliases:
KIAA1310, NSL3, Rcd1
Chromosome Location:
Chr.2: 96593155 - 96638379 on Build GRCh38
UniGene ID:
Hs.516341
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125089
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANSL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001115016.2
NP_001108488.1
15
2018
NR_047653.1
15
2079
NR_047654.1
15
2079
NR_047655.1
15
2005
NR_047656.1
14
1927
NR_047657.1
16
2096
NR_047658.1
15
1995
XM_005263982.2
XP_005264039.1
16
2095
XM_005263984.1
XP_005264041.1
16
2095
XM_005263987.1
XP_005264044.1
15
2017
XM_011511454.2
XP_011509756.1
16
2095
XM_011511455.2
XP_011509757.1
16
2064
XM_011511456.2
XP_011509758.1
16
2098
XM_011511457.2
XP_011509759.1
16
2017
XM_011511458.2
XP_011509760.1
16
2095
XM_011511459.2
XP_011509761.1
15
2020
XM_011511460.2
XP_011509762.1
15
2017
XM_011511461.2
XP_011509763.1
15
2017
XM_011511462.2
XP_011509764.1
15
1924
XM_011511463.2
XP_011509765.1
16
2108
XM_011511464.2
XP_011509766.1
16
2167
XM_011511466.2
XP_011509768.1
15
2030
XM_011511467.2
XP_011509769.1
15
1950
XM_017004470.1
XP_016859959.1
16
2095
XM_017004471.1
XP_016859960.1
15
2002
XM_017004472.1
XP_016859961.1
16
2108
XM_017004473.1
XP_016859962.1
16
2108
XM_017004474.1
XP_016859963.1
16
2003
GenBank mRNA
AB037731.1
-
16
2048
AF311326.1
-
17
1831
AK023813.1
-
15
2046
AK091163.1
-
1
1453
AK302646.1
-
15
1946
AK304011.1
-
14
1927
AK314367.1
-
15
1955
AL136849.1
-
1
19
AY050169.1
-
15
2046
BC024276.1
-
15
1955
BC051763.1
-
14
1727
BC063792.1
-
15
1948
6504387
BC127820.1
-
3
395
DB133508.1
-
2
190
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0