Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204832.1 | NP_001191761.1 | 11 | 1833 | ||
NM_001204833.1 | NP_001191762.1 | 10 | 1698 | |||
NM_001204834.1 | NP_001191763.1 | 11 | 1461 | |||
NM_018075.3 | NP_060545.3 | 12 | 2031 | |||
XM_011533885.2 | XP_011532187.2 | 14 | 2218 | |||
XM_017006716.1 | XP_016862205.1 | 13 | 2114 | |||
XM_017006717.1 | XP_016862206.1 | 13 | 2066 | |||
XM_017006718.1 | XP_016862207.1 | 13 | 2080 | |||
XM_017006719.1 | XP_016862208.1 | 12 | 1868 | |||
XM_017006721.1 | XP_016862210.1 | 11 | 1760 | |||
XM_017006722.1 | XP_016862211.1 | 11 | 1733 | |||
GenBank mRNA | AK096302.1 | - | 12 | 2031 | ||
AK131223.1 | - | 11 | 1749 | |||
AK292368.1 | - | 12 | 1916 | |||
AK295969.1 | - | 10 | 1633 | |||
AL832508.1 | - | 12 | 1898 | |||
BC038855.1 | - | 11 | 1359 |