Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017572.3 | NP_060042.2 | 6 | 611 | ||
NM_199054.2 | NP_951009.1 | 6 | 611 | |||
GenBank mRNA | AF125532.1 | - | 5 | 388 | ||
AF237775.1 | - | 5 | 402 | |||
AF237776.1 | - | 5 | 402 | |||
AK293742.1 | - | 4 | 270 | |||
AK303393.1 | - | 4 | 320 | |||
AL137565.1 | - | 2 | 73 | |||
BC073140.1 | - | 6 | 591 | 6527053 |