Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 11, 12 | 1064 | ||
XM_011511361.1 | XP_011509663.1 | 10, 11 | 818 | |||
XM_011511362.1 | XP_011509664.1 | 10, 11 | 932 | |||
XM_011511363.2 | XP_011509665.1 | 11, 12 | 1064 | |||
GenBank mRNA | AB209993.1 | - | 11, 12 | 983 | ||
AK025611.1 | - | 10, 11 | 832 | |||
AK057926.1 | - | 11, 12 | 925 | |||
AK290722.1 | - | 11, 12 | 971 | |||
AK309040.1 | - | 11, 12 | 971 | |||
BC028609.1 | - | 11, 12 | 1037 | 4826022 | ||
BC093018.1 | - | 11, 12 | 954 | 30337330 |