Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASNSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019048.2 | NP_061921.1 | 4 | 1386 | ||
XM_017004381.1 | XP_016859870.1 | 3 | 1117 | |||
XM_017004382.1 | XP_016859871.1 | 3 | 1902 | |||
GenBank mRNA | AB075481.1 | - | 1 | 835 | ||
AK000759.1 | - | 4 | 1365 | |||
AY116969.1 | - | 1 | 952 | |||
BC001243.1 | - | 4 | 1291 | 3453136 |