Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020406.3 | NP_065139.2 | 9 | 1215 | ||
XM_017027021.1 | XP_016882510.1 | 9 | 1199 | |||
XM_017027022.1 | XP_016882511.1 | 9 | 1199 | |||
GenBank mRNA | AB237911.1 | - | 9 | 1167 | ||
AF146747.1 | - | 9 | 1194 | |||
AJ290452.1 | - | 9 | 1186 | |||
AJ305326.1 | - | 9 | 1304 | |||
AJ310433.1 | - | 10 | 1305 | |||
AK225226.1 | - | 9 | 1188 | |||
AK290965.1 | - | 9 | 1190 | |||
AY358932.1 | - | 9 | 1175 | |||
BC029167.1 | - | 9 | 1177 | 5182826 | ||
BT020111.1 | - | 9 | 1159 |