Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145056.1 | NP_001138528.1 | 3 | 183 | ||
NM_020428.3 | NP_065161.3 | 3 | 242 | |||
XM_005259997.1 | XP_005260054.1 | 3 | 242 | |||
XM_005259999.1 | XP_005260056.1 | 3 | 183 | |||
GenBank mRNA | AK027519.1 | - | 3 | 127 | ||
AK314582.1 | - | 3 | 184 | |||
BC040556.1 | - | 3 | 184 | 5268313 | ||
BC042692.1 | - | 3 | 184 | |||
BI602883.1 | - | 3 | 176 | |||
DA628435.1 | - | 3 | 183 | |||
DA749803.1 | - | 3 | 242 | |||
FN668568.1 | - | 3 | 242 |