Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020890.2 | NP_065941.2 | 16 | 2190 | ||
XM_006713716.3 | XP_006713779.1 | 16 | 1974 | |||
XM_006713717.2 | XP_006713780.1 | 17 | 2086 | |||
XM_011513056.1 | XP_011511358.1 | 16 | 1934 | |||
XM_011513057.2 | XP_011511359.1 | 17 | 2055 | |||
GenBank mRNA | AB040957.1 | - | 16 | 1917 | ||
AF334474.1 | - | 16 | 1989 | |||
AK022912.1 | - | 7 | 801 | |||
AK056933.1 | - | 16 | 1999 | |||
AK308315.1 | - | 16 | 1936 | |||
AL713689.1 | - | 16 | 1985 | |||
BC017659.1 | - | 2 | 63 | |||
BC063433.1 | - | 16 | 1981 | |||
BC082961.1 | - | 16 | 1943 | |||
BC130564.1 | - | 16 | 1957 | 40146595 | ||
BC136371.1 | - | 16 | 1957 | 9020358 |