Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021156.3 | NP_066979.2 | 8 | 1265 | ||
GenBank mRNA | AB032988.1 | - | 1 | 348 | ||
AK074838.1 | - | 8 | 1197 | |||
AK075404.1 | - | 8 | 1244 | |||
AK226005.1 | - | 7 | 799 | |||
AL118494.1 | - | 7 | 790 | |||
AY359004.1 | - | 8 | 1182 | |||
BC033787.1 | - | 8 | 1076 | 5165952 | ||
BC044777.1 | - | 8 | 1265 |