Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134432.1 | NP_001127904.1 | 3 | 813 | ||
NM_001134433.1 | NP_001127905.1 | 3 | 813 | |||
NM_001271650.1 | NP_001258579.1 | 4 | 877 | |||
NM_022461.4 | NP_071906.1 | 3 | 813 | |||
XM_005265392.2 | XP_005265449.1 | 4 | 900 | |||
XM_011534009.2 | XP_011532311.1 | 3 | 353 | |||
XM_011534010.2 | XP_011532312.1 | 3 | 311 | |||
XM_011534011.2 | XP_011532313.1 | 3 | 310 | |||
GenBank mRNA | AK025592.1 | - | 3 | 660 | ||
AK290637.1 | - | 3 | 532 | |||
AY151386.1 | - | 2 | 281 | |||
BM546093.1 | - | 2 | 149 | |||
BX386357.2 | - | 3 | 734 | |||
BX648471.1 | - | 4 | 541 | |||
CB995808.1 | - | 3 | 341 |