Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320147.1 | NP_001307076.1 | 12 | 1828 | ||
NM_024046.4 | NP_076951.2 | 11 | 1921 | |||
GenBank mRNA | AK074856.1 | - | 11 | 1635 | ||
AK074899.1 | - | 12 | 1512 | |||
AK075294.1 | - | 12 | 1523 | |||
AK226073.1 | - | 3 | 672 | |||
AK295588.1 | - | 12 | 1386 | |||
AK297258.1 | - | 11 | 1567 | |||
AK297437.1 | - | 12 | 1468 | |||
AK315525.1 | - | 11 | 1654 | |||
AL136697.1 | - | 10 | 1520 | |||
AL833943.1 | - | 9 | 1979 | |||
BC000497.2 | - | 11 | 1590 | 2820734 | ||
BC001921.1 | - | 11 | 1590 | 2820734 | ||
BC005828.2 | - | 11 | 1590 | 2820734 | ||
BC017363.2 | - | 11 | 1592 | 3940018 | ||
BC019256.1 | - | 11 | 1585 | 3502894 | ||
BX647074.1 | - | 10 | 1763 | |||
CR533508.1 | - | 9 | 1358 |