Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP70 SIRNA Assays ›
Gene Symbol:
CEP70
Gene Name:
centrosomal protein 70
Gene Aliases:
BITE
Chromosome Location:
Chr.3: 138494339 - 138594383 on Build GRCh38
UniGene ID:
Hs.531962
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125443
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP70
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288964.1
NP_001275893.1
12
1283
NM_001288965.1
NP_001275894.1
11
1227
NM_001288966.1
NP_001275895.1
12
1301
NM_001288967.1
NP_001275896.1
8
831
NM_001320598.1
NP_001307527.1
12
1353
NM_001320599.1
NP_001307528.1
12
1301
NM_024491.3
NP_077817.2
12
1301
XM_005247805.3
XP_005247862.1
10
1021
XM_011513209.2
XP_011511511.1
10
1021
XM_017007270.1
XP_016862759.1
10
1021
XM_017007271.1
XP_016862760.1
12
1203
XM_017007272.1
XP_016862761.1
12
1541
XM_017007273.1
XP_016862762.1
11
1146
XM_017007274.1
XP_016862763.1
11
1566
XM_017007275.1
XP_016862764.1
12
1637
XM_017007276.1
XP_016862765.1
8
834
XM_017007277.1
XP_016862766.1
8
766
XM_017007278.1
XP_016862767.1
13
1372
XM_017007279.1
XP_016862768.1
11
1135
XM_017007280.1
XP_016862769.1
10
976
XM_017007281.1
XP_016862770.1
5
1149
XM_017007282.1
XP_016862771.1
11
1102
XM_017007283.1
XP_016862772.1
11
1033
XM_017007284.1
XP_016862773.1
5
1337
GenBank mRNA
AF202146.1
-
12
1155
AF289495.1
-
12
1155
AK023098.1
-
10
1016
AK225440.1
-
12
1166
AK294587.1
-
8
706
AK299003.1
-
11
1092
AK301913.1
-
12
1160
AK314314.1
-
12
1273
BC030598.2
-
12
1203
4829481
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0