Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARMC9 SIRNA Assays ›
Gene Symbol:
ARMC9
Gene Name:
armadillo repeat containing 9
Gene Aliases:
ARM, KU-MEL-1, NS21
Chromosome Location:
Chr.2: 231198546 - 231394991 on Build GRCh38
UniGene ID:
Hs.162411
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125599
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARMC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271466.2
NP_001258395.1
16
1604
NM_001291656.1
NP_001278585.1
17
1757
NM_025139.5
NP_079415.3
17
1759
XM_011511905.1
XP_011510207.1
17
1677
XM_011511906.1
XP_011510208.1
17
1677
XM_011511907.1
XP_011510209.1
16
1578
XM_011511908.2
XP_011510210.1
17
1677
XM_011511909.2
XP_011510211.1
17
1677
XM_011511910.1
XP_011510212.1
17
1677
XM_011511911.1
XP_011510213.1
17
1677
XM_011511912.1
XP_011510214.1
17
1677
XM_011511913.2
XP_011510215.1
17
1677
XM_011511914.2
XP_011510216.1
17
1677
XM_011511915.2
XP_011510217.1
17
1677
XM_011511916.2
XP_011510218.1
17
1677
XM_011511919.2
XP_011510221.1
7
1065
XM_017005018.1
XP_016860507.1
17
1935
XM_017005019.1
XP_016860508.1
17
1853
XM_017005020.1
XP_016860509.1
17
1935
XM_017005021.1
XP_016860510.1
17
1677
XM_017005022.1
XP_016860511.1
17
1677
XM_017005023.1
XP_016860512.1
17
1935
XM_017005024.1
XP_016860513.1
16
1578
XM_017005025.1
XP_016860514.1
17
1677
XM_017005026.1
XP_016860515.1
17
1876
GenBank mRNA
AB058771.1
-
6
500
AK022646.1
-
15
1461
AY219922.1
-
17
1638
AY929062.1
-
17
1631
BC004514.2
-
13
1181
BC065271.1
-
16
1591
6165433
BX537956.1
-
17
1675
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-k7c2b:80/10.239.160.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0