Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318226.1 | NP_001305155.1 | 7 | 935 | ||
NM_025176.5 | NP_079452.3 | 7 | 935 | |||
XM_011529186.2 | XP_011527488.1 | 7 | 1558 | |||
XM_011529187.2 | XP_011527489.1 | 7 | 1558 | |||
XM_011529188.1 | XP_011527490.1 | 7 | 935 | |||
XM_011529189.1 | XP_011527491.1 | 7 | 943 | |||
XM_011529190.1 | XP_011527492.1 | 6 | 858 | |||
XM_011529191.1 | XP_011527493.1 | 3 | 450 | |||
XM_011529193.1 | XP_011527495.1 | 6 | 858 | |||
XM_017027732.1 | XP_016883221.1 | 8 | 2877 | |||
XM_017027733.1 | XP_016883222.1 | 8 | 1004 | |||
GenBank mRNA | AB023197.1 | - | 7 | 921 | ||
AK293991.1 | - | 7 | 1221 | |||
BC036380.2 | - | 6 | 873 | 5268454 | ||
EU718622.1 | - | 7 | 921 |