Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM49A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030797.3 | NP_110424.1 | 11 | 1105 | ||
XM_005262630.3 | XP_005262687.1 | 10 | 1061 | |||
XM_006712108.3 | XP_006712171.1 | 11 | 1060 | |||
GenBank mRNA | AK001942.1 | - | 3 | 161 | ||
AK055334.1 | - | 11 | 1073 | |||
AK091280.1 | - | 6 | 519 | |||
AK315065.1 | - | 10 | 1000 | |||
AL136704.1 | - | 11 | 1070 | |||
BC038971.1 | - | 11 | 1056 | 5200807 | ||
CR533504.1 | - | 9 | 852 |