Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM49A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030797.3 | NP_110424.1 | 12 | 1191 | ||
XM_005262630.3 | XP_005262687.1 | 11 | 1147 | |||
XM_006712108.3 | XP_006712171.1 | 12 | 1146 | |||
GenBank mRNA | AK001942.1 | - | 4 | 247 | ||
AK055334.1 | - | 12 | 1159 | |||
AK091280.1 | - | 7 | 605 | |||
AL136704.1 | - | 12 | 1156 | |||
BC038971.1 | - | 12 | 1142 | 5200807 | ||
CR533504.1 | - | 10 | 938 |