Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030915.3 | NP_112177.2 | 3 | 1863 | ||
XM_017005059.1 | XP_016860548.1 | 3 | 2021 | |||
GenBank mRNA | AF110224.1 | - | 1 | 632 | ||
AF258555.1 | - | 1 | 854 | |||
AK023425.1 | - | 1 | 930 | |||
AK027232.1 | - | 3 | 1655 | |||
AK223541.1 | - | 3 | 1799 | |||
AL136703.1 | - | 3 | 1868 | |||
BC109376.1 | - | 3 | 1867 | 6495944 | ||
BC126434.1 | - | 3 | 1836 | 8992150 | ||
BC130415.1 | - | 3 | 1835 | 40146446 | ||
EF025586.1 | - | 3 | 1841 |