Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289177.1 | NP_001276106.1 | 3 | 414 | ||
NM_001289178.1 | NP_001276107.1 | 3 | 360 | |||
NM_001289179.1 | NP_001276108.1 | 3 | 405 | |||
NM_001289180.1 | NP_001276109.1 | 4 | 245 | |||
NM_001321384.1 | NP_001308313.1 | 3 | 395 | |||
NM_030924.4 | NP_112186.3 | 3 | 405 | |||
XM_005259653.3 | XP_005259710.1 | 3 | 387 | |||
XM_017027332.1 | XP_016882821.1 | 3 | 232 | |||
XM_017027333.1 | XP_016882822.1 | 3 | 387 | |||
XM_017027334.1 | XP_016882823.1 | 3 | 387 | |||
GenBank mRNA | AJ577571.1 | - | 3 | 308 | ||
AK093433.1 | - | 3 | 373 | |||
AK225862.1 | - | 2 | 113 | |||
AK302604.1 | - | 3 | 362 | |||
AL136854.1 | - | 3 | 146 | |||
AY009107.1 | - | 4 | 238 | |||
AY358766.1 | - | 3 | 405 | |||
BC022027.1 | - | 3 | 336 | 4825681 | ||
BC090046.1 | - | 3 | 321 | 30388900 | ||
BI561348.1 | - | 3 | 366 | |||
DC399703.1 | - | 4 | 245 | |||
HY011431.1 | - | 3 | 392 |