Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005474.2 | NP_001005474.1 | 13 | 3698 | ||
NM_031419.3 | NP_113607.1 | 12 | 3831 | |||
GenBank mRNA | AB037925.1 | - | 12 | 3763 | ||
AF548362.1 | - | 13 | 3694 | |||
BC014587.2 | - | 3747 | ||||
BC060800.1 | - | 12 | 3830 | 30344780 |