Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184957.1 | NP_001171886.1 | 12 | 1743 | ||
NM_001184958.1 | NP_001171887.1 | 12 | 1848 | |||
NM_032137.4 | NP_115513.4 | 12 | 2245 | |||
XM_006713341.2 | XP_006713404.1 | 12 | 2245 | |||
XM_006713342.2 | XP_006713405.1 | 12 | 2098 | |||
XM_006713343.2 | XP_006713406.1 | 12 | 2245 | |||
XM_011534153.2 | XP_011532455.1 | 12 | 3875 | |||
XM_011534154.2 | XP_011532456.1 | 12 | 3310 | |||
XM_011534157.2 | XP_011532459.1 | 8 | 1022 | |||
XM_011534158.2 | XP_011532460.1 | 11 | 2121 | |||
GenBank mRNA | AK131481.1 | - | 13 | 2451 | ||
AL136781.1 | - | 12 | 1663 | |||
AL834386.1 | - | 12 | 2197 | |||
BC030599.2 | - | 12 | 1806 | 4831210 | ||
BC038406.1 | - | 12 | 2165 | 5167835 |