Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303096.1 | NP_001290025.1 | 10 | 951 | ||
NM_032168.2 | NP_115544.1 | 9 | 917 | |||
GenBank mRNA | AK022581.1 | - | 9 | 864 | ||
AK022607.1 | - | 9 | 876 | |||
AK023276.1 | - | 6 | 556 | |||
AK091546.1 | - | 10 | 878 | |||
AK290445.1 | - | 9 | 868 | |||
AL122097.1 | - | 5 | 484 | |||
BC040567.1 | - | 9 | 917 | 5267120 | ||
CR749440.1 | - | 9 | 2526 |