Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032242.3 | NP_115618.3 | 14 | 2889 | ||
XM_011512908.2 | XP_011511210.1 | 15 | 2991 | |||
XM_011512909.1 | XP_011511211.1 | 15 | 3584 | |||
GenBank mRNA | X87832.2 | - | 14 | 2824 |