Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011537.2 | NP_001011537.2 | 10 | 1282 | ||
NM_032288.6 | NP_115664.2 | 9 | 1341 | |||
NR_027840.1 | 9 | 1452 | ||||
GenBank mRNA | AJ344094.1 | - | 9 | 1187 | ||
AK027672.1 | - | 9 | 1225 | |||
AK316308.1 | - | 9 | 1082 | |||
AL136558.1 | - | 9 | 1195 | |||
BC035006.2 | - | 9 | 1344 | 5267824 | ||
BC039734.1 | - | 9 | 1249 | 6042926 |