Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256421.1 | NP_001243350.1 | 4 | 584 | ||
NM_001256422.1 | NP_001243351.1 | 4 | 588 | |||
NM_032717.4 | NP_116106.2 | 3 | 671 | |||
XM_011532383.1 | XP_011530685.1 | 2 | 477 | |||
XM_011532384.2 | XP_011530686.1 | 4 | 2672 | |||
XM_017008780.1 | XP_016864269.1 | 4 | 740 | |||
XM_017008781.1 | XP_016864270.1 | 4 | 747 | |||
GenBank mRNA | AK055749.1 | - | 4 | 583 | ||
AK223543.1 | - | 4 | 555 | |||
AY358100.1 | - | 3 | 488 | |||
BC090956.1 | - | 3 | 577 | 30387839 | ||
DQ324782.1 | - | 3 | 368 | |||
DQ345298.1 | - | 3 | 668 |