Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322080.1 | NP_001309009.1 | 6 | 2347 | ||
NM_001322081.1 | NP_001309010.1 | 6 | 2248 | |||
NM_032718.4 | NP_116107.3 | 6 | 2231 | |||
NR_136173.1 | 7 | 2315 | ||||
XM_011512005.2 | XP_011510307.1 | 6 | 2354 | |||
XM_011512006.2 | XP_011510308.1 | 6 | 2171 | |||
XM_011512007.2 | XP_011510309.1 | 4 | 1828 | |||
GenBank mRNA | AK075376.1 | - | 6 | 2197 |