Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256476.1 | NP_001243405.1 | 1, 2 | 296 | ||
NM_138458.3 | NP_612467.1 | 1, 2 | 296 | |||
GenBank mRNA | AK056303.1 | - | 1, 2 | 260 | ||
AY541587.1 | - | 1, 2 | 271 | |||
BC014022.2 | - | 1, 2 | 218 | 4309224 | ||
BC020271.1 | - | 1, 2 | 250 | |||
BC066657.1 | - | 1, 2 | 246 | 6164689 | ||
BP271204.1 | - | 1, 2 | 296 |