Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301069.1 | NP_001287998.1 | 1 | 428 | ||
NM_003089.5 | NP_003080.2 | 1 | 428 | |||
GenBank mRNA | AK096783.1 | - | 1 | 117 | ||
AK293120.1 | - | 1 | 147 | |||
AL560281.3 | - | 1 | 115 | |||
BC001315.1 | - | 1 | 101 | 3454479 | ||
BE397125.1 | - | 1 | 299 | |||
DA403443.1 | - | 1 | 409 | |||
DB189176.1 | - | 1 | 385 | |||
M22636.1 | - | 1 | 92 | |||
X06815.1 | - | 1 | 73 | |||
X06817.1 | - | 1 | 136 |