Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PACRGL SIRNA Assays ›
Gene Symbol:
PACRGL
Gene Name:
PARK2 coregulated like
Gene Aliases:
C4orf28
Chromosome Location:
Chr.4: 20696282 - 20757766 on Build GRCh38
UniGene ID:
Hs.479298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126023
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PACRGL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130727.1
NP_001124199.1
5
811
NM_001258345.1
NP_001245274.1
9
1186
NM_001258346.1
NP_001245275.1
7
879
NM_001317849.1
NP_001304778.1
6
906
NM_145048.3
NP_659485.1
8
1105
NR_047661.1
8
919
XM_011513780.1
XP_011512082.1
12
1491
XM_011513781.1
XP_011512083.1
12
1310
XM_011513784.1
XP_011512086.1
12
1327
XM_011513785.1
XP_011512087.1
11
1410
XM_011513786.2
XP_011512088.1
11
1349
XM_011513787.1
XP_011512089.1
10
1282
XM_011513788.1
XP_011512090.1
11
1399
XM_011513791.1
XP_011512093.1
10
1242
XM_011513792.1
XP_011512094.1
10
1237
XM_011513794.1
XP_011512096.1
11
1190
XM_011513795.1
XP_011512097.1
10
1112
XM_011513796.1
XP_011512098.1
10
1107
XM_011513797.2
XP_011512099.1
10
1113
XM_011513798.2
XP_011512100.1
10
1049
XM_011513804.1
XP_011512106.1
8
915
XM_011513805.2
XP_011512107.1
8
890
XM_017007747.1
XP_016863236.1
10
1104
XM_017007748.1
XP_016863237.1
10
1055
XM_017007749.1
XP_016863238.1
9
970
XM_017007750.1
XP_016863239.1
9
1240
XM_017007751.1
XP_016863240.1
9
1172
XM_017007752.1
XP_016863241.1
8
1019
XM_017007753.1
XP_016863242.1
8
889
XM_017007754.1
XP_016863243.1
6
946
XM_017007755.1
XP_016863244.1
6
763
XM_017007756.1
XP_016863245.1
6
756
XM_017007757.1
XP_016863246.1
6
692
XM_017007758.1
XP_016863247.1
5
641
XM_017007759.1
XP_016863248.1
5
589
GenBank mRNA
AK291544.1
-
9
1142
AK294077.1
-
7
834
AK297556.1
-
5
622
AK307355.1
-
8
874
BC023002.2
-
8
852
4992139
BG741791.1
-
4
421
BX344401.2
-
6
841
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline