Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002986.2 | NP_002977.1 | 3 | 543 | ||
GenBank mRNA | U46573.1 | - | 3 | 455 | ||
Z75668.1 | - | 4 | 457 |