Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152672.5 | NP_689885.4 | 9 | 1256 | ||
GenBank mRNA | AK172837.1 | - | 9 | 1259 | ||
BC029606.1 | - | 5 | 868 | 5297443 |