Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126204
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
7
785
NM_001283116.2
NP_001270045.1
6
539
NM_001283129.2
NP_001270058.1
6
779
NM_001283148.2
NP_001270077.1
6
539
NM_001283154.2
NP_001270083.1
6
651
NM_001283215.2
NP_001270144.1
6
779
NM_001283235.2
NP_001270164.1
5
424
NM_001322141.1
NP_001309070.1
6
779
NM_001322142.1
NP_001309071.1
6
539
NM_001322143.1
NP_001309072.1
6
852
NM_001322144.1
NP_001309073.1
6
779
NM_001322146.1
NP_001309075.1
6
689
NM_001322148.1
NP_001309077.1
6
856
NM_001322150.1
NP_001309079.1
5
664
NM_001322153.1
NP_001309082.1
5
424
NM_001322155.1
NP_001309084.1
5
497
NM_001322160.1
NP_001309089.1
6
741
NM_001322171.1
NP_001309100.1
5
497
NM_001322172.1
NP_001309101.1
5
789
NM_001322173.1
NP_001309102.1
5
664
NM_001322174.1
NP_001309103.1
5
551
NM_001322175.1
NP_001309104.1
5
549
NM_152906.6
NP_690870.3
6
651
NR_104274.2
5
424
NR_104275.2
5
664
NR_136211.1
6
612
NR_136212.1
5
497
XM_011529863.1
XP_011528165.1
6
690
XM_011529865.1
XP_011528167.1
5
555
XM_011529867.1
XP_011528169.1
6
634
XM_017028577.1
XP_016884066.1
6
612
XM_017028578.1
XP_016884067.1
6
631
XM_017028579.1
XP_016884068.1
6
660
XM_017028580.1
XP_016884069.1
7
749
XM_017028581.1
XP_016884070.1
6
743
XM_017028582.1
XP_016884071.1
7
826
XM_017028583.1
XP_016884072.1
6
573
XM_017028584.1
XP_016884073.1
6
836
XM_017028587.1
XP_016884076.1
6
743
XM_017028588.1
XP_016884077.1
6
592
GenBank mRNA
AK289862.1
-
7
754
AK295210.1
-
5
665
AK297922.1
-
6
497
AK298593.1
-
5
382
AK301054.1
-
5
383
AK301366.1
-
6
738
AK311676.1
-
5
383
AK316020.1
-
6
500
AK316056.1
-
6
759
AL713640.1
-
3
279
BC041339.1
-
6
614
5284126
BF726547.1
-
2
136
BG685068.1
-
4
389
BI544307.1
-
6
614
BM048105.1
-
120
BP303570.1
-
5
436
BU173566.1
-
6
571
CD675132.1
-
5
478
CF552364.1
-
5
510
CR456355.1
-
5
466
CR627452.1
-
5
2886
DB021425.1
-
4
329
DC301369.1
-
5
393
DC315148.1
-
5
382
DC318353.1
-
5
424
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline