Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039573.2 | NP_001034662.2 | 6, 7 | 803 | ||
NM_001143998.1 | NP_001137470.1 | 6, 7 | 803 | |||
NM_001143999.1 | NP_001137471.1 | 6, 7 | 799 | |||
NM_001144001.1 | NP_001137473.1 | 4, 5 | 591 | |||
NM_001204408.1 | NP_001191337.1 | 8, 9 | 1148 | |||
NM_001204410.1 | NP_001191339.1 | 7, 8 | 841 | |||
NM_003003.3 | NP_002994.3 | 6, 7 | 750 | |||
GenBank mRNA | AB208811.1 | - | 6, 7 | 782 | ||
AK290913.1 | - | 6, 7 | 696 | |||
AK293206.1 | - | 4, 5 | 591 | |||
BC136523.1 | - | 6, 7 | 638 | 9020513 | ||
BC136525.1 | - | 6, 7 | 638 | 9020515 | ||
BC142979.1 | - | 6, 7 | 694 | 8860414 | ||
BM802278.1 | - | 324 | ||||
BX537448.1 | - | 6, 7 | 704 | |||
D67029.1 | - | 6, 7 | 786 | |||
FM995495.1 | - | 8, 9 | 1042 | |||
FM995496.1 | - | 7, 8 | 839 | |||
FM995497.1 | - | 7, 8 | 938 |