Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 26 | 3851 | ||
XM_005246299.4 | XP_005246356.1 | 27 | 3929 | |||
XM_005246300.4 | XP_005246357.1 | 27 | 3884 | |||
XM_006712268.3 | XP_006712331.1 | 27 | 3926 | |||
XM_006712269.3 | XP_006712332.1 | 26 | 3851 | |||
XM_006712270.3 | XP_006712333.2 | 26 | 3785 | |||
XM_006712271.3 | XP_006712334.1 | 26 | 3892 | |||
XM_011510604.2 | XP_011508906.1 | 27 | 3884 | |||
XM_017003356.1 | XP_016858845.1 | 27 | 3884 | |||
XM_017003357.1 | XP_016858846.1 | 27 | 3884 | |||
XM_017003358.1 | XP_016858847.1 | 27 | 3884 | |||
GenBank mRNA | AK126998.1 | - | 4 | 580 | ||
AK128490.1 | - | 10 | 1331 | |||
BX649036.1 | - | 2 | 87 | |||
CR749469.1 | - | 4 | 341 |