Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 39 | 5987 | ||
XM_005246299.4 | XP_005246356.1 | 40 | 6077 | |||
XM_005246300.4 | XP_005246357.1 | 40 | 6020 | |||
XM_006712268.3 | XP_006712331.1 | 40 | 6074 | |||
XM_006712269.3 | XP_006712332.1 | 39 | 5999 | |||
XM_006712270.3 | XP_006712333.2 | 39 | 5933 | |||
XM_006712271.3 | XP_006712334.1 | 39 | 6040 | |||
GenBank mRNA | AB095944.1 | - | 10 | 2096 | ||
AK094741.1 | - | 3 | 759 | |||
AK126998.1 | - | 17 | 2716 | |||
AK127553.1 | - | 13 | 1994 | |||
AK128490.1 | - | 23 | 3467 | |||
AK303062.1 | - | 14 | 2087 | |||
AL834144.1 | - | 11 | 1694 | |||
BC044594.1 | - | 1 | 405 | |||
BX494687.1 | - | 1 | 260 | |||
BX649036.1 | - | 15 | 2223 | |||
CR749469.1 | - | 17 | 2488 |