Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130726.2 | NP_001124198.1 | 5, 6 | 581 | ||
NM_173463.4 | NP_775734.2 | 6, 7 | 682 | |||
XM_005248210.3 | XP_005248267.1 | 5, 6 | 610 | |||
XM_011513906.2 | XP_011512208.1 | 7, 8 | 692 | |||
XM_011513907.2 | XP_011512209.1 | 6, 7 | 565 | |||
XM_011513908.1 | XP_011512210.1 | 5, 6 | 487 | |||
XM_017008827.1 | XP_016864316.1 | 7, 8 | 692 | |||
XM_017008828.1 | XP_016864317.1 | 6, 7 | 616 | |||
GenBank mRNA | AK125353.1 | - | 5, 6 | 490 | ||
AK296454.1 | - | 2, 3 | 239 | |||
AK302905.1 | - | 6, 7 | 626 | |||
AL834257.2 | - | 5, 6 | 555 | |||
DR423861.1 | - | 5, 6 | 496 |