Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291419.1 | NP_001278348.1 | 4 | 693 | ||
NM_173506.6 | NP_775777.3 | 4 | 1668 | |||
XM_005258543.3 | XP_005258600.1 | 4 | 696 | |||
XM_005278383.4 | XP_005278440.1 | 4 | 1707 | |||
XM_005278385.4 | XP_005278442.1 | 4 | 1707 | |||
XM_005278387.4 | XP_005278444.1 | 4 | 1707 | |||
XM_005278388.3 | XP_005278445.1 | 3 | 1491 | |||
XM_006723035.2 | XP_006723098.1 | 3 | 484 | |||
XM_011526483.2 | XP_011524785.1 | 4 | 864 | |||
XM_011526484.2 | XP_011524786.1 | 4 | 769 | |||
GenBank mRNA | AK292305.1 | - | 4 | 1663 | ||
AY358726.1 | - | 4 | 1437 | |||
AY771680.1 | - | 3 | 454 | |||
BC034629.2 | - | 4 | 693 | 4827584 |