Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284263.1 | NP_001271192.1 | 4 | 1069 | ||
NM_001284264.1 | NP_001271193.1 | 4 | 898 | |||
NM_001284265.1 | NP_001271194.1 | 5 | 849 | |||
NM_173510.3 | NP_775781.1 | 5 | 1123 | |||
GenBank mRNA | AK091133.1 | - | 5 | 1049 | ||
AK127794.1 | - | 1 | 1340 | |||
AK289516.1 | - | 5 | 1121 | |||
AK295140.1 | - | 4 | 798 | |||
AK302798.1 | - | 5 | 852 | |||
AK302925.1 | - | 4 | 1015 | |||
BC040488.1 | - | 5 | 1078 | 5272169 | ||
BC053874.1 | - | 5 | 1083 | 5531689 | ||
BC065549.1 | - | 5 | 1085 | 6045091 | ||
CR456461.1 | - | 4 | 646 |