Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284263.1 | NP_001271192.1 | 4 | 1377 | ||
NM_001284264.1 | NP_001271193.1 | 4 | 1206 | |||
NM_001284265.1 | NP_001271194.1 | 5 | 1157 | |||
NM_173510.3 | NP_775781.1 | 5 | 1431 | |||
GenBank mRNA | AK091133.1 | - | 5 | 1357 | ||
AK127794.1 | - | 1 | 1648 | |||
AK289516.1 | - | 5 | 1429 | |||
AK295140.1 | - | 4 | 1106 | |||
BC040488.1 | - | 5 | 1386 | 5272169 | ||
BC053874.1 | - | 5 | 1391 | 5531689 |