Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITSN1 SIRNA Assays ›
Gene Symbol:
ITSN1
Gene Name:
intersectin 1
Gene Aliases:
ITSN, SH3D1A, SH3P17
Chromosome Location:
Chr.21: 33642400 - 33899861 on Build GRCh38
UniGene ID:
Hs.160324
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
12635
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ITSN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001132.1
NP_001001132.1
8
953
NM_003024.2
NP_003015.2
8
953
XM_005261026.2
XP_005261083.1
8
1031
XM_005261027.1
XP_005261084.1
8
1031
XM_005261031.1
XP_005261088.1
8
1031
XM_011529691.1
XP_011527993.1
8
920
XM_011529692.1
XP_011527994.1
8
920
XM_011529693.2
XP_011527995.1
8
1031
XM_017028428.1
XP_016883917.1
9
951
XM_017028429.1
XP_016883918.1
8
920
XM_017028430.1
XP_016883919.1
8
920
XM_017028431.1
XP_016883920.1
8
1031
XM_017028432.1
XP_016883921.1
8
1031
XM_017028433.1
XP_016883922.1
8
1031
XM_017028434.1
XP_016883923.1
8
1031
XM_017028435.1
XP_016883924.1
8
920
XM_017028436.1
XP_016883925.1
8
920
XM_017028437.1
XP_016883926.1
4
487
XM_017028438.1
XP_016883927.1
8
920
XM_017028439.1
XP_016883928.1
8
1031
XM_017028440.1
XP_016883929.1
8
920
XM_017028441.1
XP_016883930.1
8
1031
GenBank mRNA
AF064243.1
-
8
791
AF064244.1
-
8
791
AF114487.1
-
8
953
AF114488.1
-
8
953
AK027846.1
-
8
920
AK074554.1
-
8
889
AK075290.1
-
8
948
AK300274.1
-
8
928
BC013578.1
-
8
816
BC020269.1
-
8
866
BC039036.1
-
8
948
BC058925.1
-
8
980
BC073744.1
-
8
895
BC116185.1
-
8
764
40073780
BC116186.2
-
8
764
40073781
BC117560.1
-
8
933
7469144
BX538175.1
-
3
252
DQ340865.1
-
7
681
DQ386455.1
-
7
704
DQ679754.1
-
8
770
DQ679755.1
-
8
770
DQ679756.1
-
9
659
DQ679757.1
-
8
770
EU107386.1
-
8
659
EU117382.1
-
7
686
EU120733.1
-
7
686
EU120734.1
-
7
686
EU120735.1
-
8
659
EU120736.1
-
8
659
EU140799.1
-
8
770
EU140800.1
-
7
575
EU152331.1
-
7
686
EU191901.1
-
7
609
GU939616.1
-
3
375
GU939617.1
-
3
375
GU939621.1
-
3
677
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
regulation of Rho protein signal transduction
positive regulation of phosphorylation
positive regulation of apoptotic process
negative regulation of neuron apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
synaptic vesicle endocytosis
regulation of small GTPase mediated signal transduction
positive regulation of protein kinase B signaling
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
calcium ion binding
protein binding
kinase activator activity
protein complex scaffold
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0