Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040045.1 | 3 | 834 | |||
GenBank mRNA | AK092743.1 | - | 3 | 832 | ||
AX747702.1 | - | 3 | 832 |