Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031749.2 | NP_001026919.2 | 5 | 1231 | ||
NM_001288763.1 | NP_001275692.1 | 4 | 1281 | |||
NM_182573.2 | NP_872379.2 | 5 | 1203 | |||
XM_011526794.2 | XP_011525096.1 | 5 | 1287 | |||
GenBank mRNA | AK055031.1 | - | 5 | 1181 | ||
BC057816.1 | - | 5 | 1217 | 30345834 |