Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115007.2 | NP_001108479.1 | 8 | 923 | ||
NM_001115008.2 | NP_001108480.1 | 8 | 1124 | |||
NM_001288996.1 | NP_001275925.1 | 5 | 1520 | |||
NM_001288997.1 | NP_001275926.1 | 8 | 861 | |||
NM_194282.3 | NP_919258.2 | 8 | 1787 | |||
NR_110253.1 | 5 | 1446 | ||||
NR_110254.1 | 7 | 1120 | ||||
XM_005262750.4 | XP_005262807.1 | 7 | 1596 | |||
XM_006714081.3 | XP_006714144.1 | 7 | 1522 | |||
XM_017007728.1 | XP_016863217.1 | 5 | 1329 | |||
XM_017007729.1 | XP_016863218.1 | 6 | 1512 | |||
XM_017007730.1 | XP_016863219.1 | 6 | 901 | |||
GenBank mRNA | AB111889.1 | - | 7 | 1107 | ||
AK292769.1 | - | 8 | 871 | |||
AK294256.1 | - | 8 | 1716 | |||
AK299787.1 | - | 3 | 329 | |||
BC109277.2 | - | 5 | 1322 | |||
BC109278.2 | - | 5 | 1395 | 40028371 | ||
BX537919.1 | - | 7 | 1021 | |||
BX640657.1 | - | 8 | 850 | |||
BX640966.1 | - | 8 | 1782 |