Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198563.2 | NP_940965.1 | 1, 2 | 288 | ||
GenBank mRNA | AY726591.1 | - | 1, 2 | 52 | ||
BC047015.1 | - | 1, 2 | 238 | 5199769 | ||
BX360795.2 | - | 1, 2 | 303 |
Gene Symbol: TMEM110-MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198974.2 | NP_001185903.2 | 1, 2 | 288 |